Pierce TMT™ Elution Buffer
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90104
PIER90104EA 113 EUR InStock
PIER90104
Pierce TMT™ Elution Buffer
Puffer Assay-Puffer
Thermo Scientific TMT Elution Buffer is for use with Thermo Scientific Immobilized Anti-TMT Resin to enrich samples for peptides that have been chemically labeled with Tandem Mass Tag™ (TMT™) Reagents.
  • For use with a mass spectrometer
  • Designed to enrich chemically labeled samples for peptides
  • Helps provide higher specific enrichment compared to acidic or basic elution buffers
Immobilized Anti-TMT Resin and TMT Elution Buffer are effective for reducing sample complexity, improving dynamic range and studying cysteine modifications such as S-nitrosylation, oxidation and disulfide bonds of protein samples labeled with iodoTMT Reagents. This approach of selective thiol labeling and affinity enrichment is similar to isotope-coded affinity tags (ICAT™ Method) but allows for higher multiplex quantitation.
Achtung! Bitte beachten: For Research Use Only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge