Mass tagging, mass tag labelling reagents, AminoxyTMT™, Pierce™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90401 90402
PIER90401EA 748 EUR InStock
PIER90401 PIER90402
Mass tagging, mass tag labelling reagents, AminoxyTMT™, Pierce™
Protein-Extraktion Proteinextraktions-Reagenzien
The carbonyl-reactive Thermo Scientific™ aminoxyTMT™ (Tandem Mass Tag™) Label Reagents enable multiplexed characterisation and quantitation of carbonyl-containing biomolecules (carbohydrates, steroids, oxidised proteins) by mass spectrometry (MS).
  • Achieve labelling efficiency greater than 90% in one hour
  • Quantitative and stable
  • 20-fold increase in signal-to-noise compared to unlabelled native glycan MS analysis
  • Optimised for excellent labelling efficiency and recovery of glycans
Native glycans are difficult to study by mass spectrometry because of their poor ionisation efficiency. Quantitation of glycans is particularly challenging due to the lack of standards for all naturally-occurring glycans and difficulties reproducibly quantifying multiple samples. The aminoxy group has better reactivity with carbonyls and better stability of the labeled product compared to hydrasides. Labelling with the AminoxyTMT™ reagents improves ionisation of glycans, thus improving sensitivity, and enables relative quantitation of glycans for up to six samples concurrently.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge