Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Premium-Qualität, Pierce™

Lieferant: Thermo Scientific

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

PG82085 PG82086
PIERPG82085EA 628 EUR InStock
PIERPG82085 PIERPG82086
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Premium-Qualität, Pierce™
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Thermo Scientific Pierce Premium Grade Sulfo-SMCC is our highest quality formulation of amine-to-sulfhydryl crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.

  • Sulfo-SMCC—a water-soluble amine-to-sulfhydryl crosslinker with a stabilizing cyclohexane spacer arm
  • High quality—identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review

Compared to the standard grade product, Premium Grade Sulfo-SMCC provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-SMCC is a water-soluble, non-cleavable and membrane-impermeable amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 angstroms). NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of Sulfo-SMCC are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm. Because it contains the hydrophilic sulfonyl moiety, Sulfo-SMCC is soluble up to nearly 10 mM in water and many commonly used buffers, thus avoiding the use of organic solvents which may perturb protein structure.

Achtung! Bitte beachten: For Research Use Only. Not for use in diagnostic procedures.


Formel: C₁₆H₁₈N₂O₉S
Storage Temperature: Gefrierschrank
MDL Number: MFCD00054978
CAS-Nummer: 92921-24-9
Order Now
Spezifikation Testergebnisse
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Short
Form Powder
Quantity 100 mg/1 g
Type Crosslinker
Solubility Water

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge