Laboratory Pipetting Needles with Blunt Ends
Lieferant: CADENCE SCIENCE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7956
POPR7956EA 2460 EUR InStock
POPR7956
Laboratory Pipetting Needles with Blunt Ends
Injektionsnadeln
All needles are made with type 304 stainless steel cannulae. Needles fitted with stainless steel hubs are designed for special applications with corrosive materials or critical procedures. Needles fitted with standard nickel plated Luer hubs are used in combination with both manual and automatic volumetric dispensers, dilutors and pipettors.
Micro-Mate® pipetting needles are developed for those applications requiring the use of plastic syringes with pipetting needles. These 90° blunt end needles are made with a 316 stainless steel cannula and a special 316 stainless Micro-Mate® female Luer hub. These hubs provide a secure, leakproof mating of plastic Luer tip syringes with metal Luer pipetting needles.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge