Glass fibre filters, without binder
Lieferant: PRAT DUMAS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A0258837
PRATA0258837EA 50.9 EUR InStock
PRATA0258837
Glass fibre filters, without binder
Filter Glasfaserfilter
Borosilicate glass fibre filters without binder.
  • Pure quality in white colour
  • For use in very high-temperature (380 °C)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge