Autoclaves, large capacity, Priorclave
Lieferant: Priorclave
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
EH230-SMART EH350-SMART EH450-SMART EH700-SMART EH450-BASE EH350-BASE EH700-BASE EH230-BASE EH285-BASE EH285-SMART
PRIOEH230-SMARTEA 0 EUR InStock
PRIOEH230-SMART PRIOEH350-SMART PRIOEH450-SMART PRIOEH700-SMART PRIOEH450-BASE PRIOEH350-BASE PRIOEH700-BASE PRIOEH230-BASE PRIOEH285-BASE PRIOEH285-SMART
Autoclaves, large capacity, Priorclave
Sterilisatoren Autoklaven (Dampfsterilisatoren)
These autoclaves combine a huge internal capacity with a small floor area and are ideally suited for use in a wide range of sterilising applications such as media preparation, liquids and diluent, waste, glassware, instruments and apparatus. They are extensively used within many industries, including food, drink, dairy, pharmaceutical, agricultural, education, healthcare, research establishments and industry. All Priorclaves come complete with Biomaster antimicrobial coating, helping improved laboratory hygiene; it is effective against all bacteria and fungi including MRSA and thereforeMore Product Information
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..