Slide Printer, Signature
Lieferant: Primera Technology Europe
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
078108
PRMA078108EA 11160 EUR InStock
PRMA078108
Slide Printer, Signature
Drucker Cassette and Slide Printers
The Signature slide printer can significantly increase laborator efficiency while helping to reduce the risk of incorrect sample identification. It prints directly onto slides, eliminating handwriting errors and the need for difficult to apply, xylene-resistant labels.
  • Efficient, hands-free operation
  • Slides are stored in easy-to-load cartridges away from dust and other potential contaminants
  • USB interface enables integration with laboratory information systems
  • Chemical resistant ink ensures reliable identification of slides
With 300 dpi print resolution, the slide printed can be used to print text, graphics and logos along with high-resolution linear and 2D bar codes on every slide. The level of slides remaining in a cartridge is easily viewed through the cassette's transparent blue LED backlit case. Print speed: Up to 9 slides/minute (monochrome); 5 slides/minute (solid colours); 4 slides/minute (process colours).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge