Reference materials, acid number standards, total acid number (TAN)
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CON150-800-051 CON150-800-011 CON150-800-151 CON150-800-455 CON150-800-205
PROOCON150-800-051EA 0 EUR InStock
PROOCON150-800-051 PROOCON150-800-011 PROOCON150-800-151 PROOCON150-800-455 PROOCON150-800-205
Reference materials, acid number standards, total acid number (TAN)
Standards Mineralölindustrie-Standards
Acid number reference materials are intended for use in the determination of acid number (AN) in petroleum products in accordance with ASTM D664/IP 177 (by potentiometric titration) or ASTM D974 (by colour-indicator titration).
Zertifizierungen: GMP/GLP, ISO 9001, ISO/IEC 17025 and ISO/IEC 17043 certified.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge