Organic reference standard, fortified human milk - organic contaminants, NIST SRM 1954
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
NIST-1954
PROONIST-1954EA 1220 EUR InStock
PROONIST-1954
Organic reference standard, fortified human milk - organic contaminants, NIST SRM 1954
Standards Organische Standards
This reference standard intended for use in evaluating analytical methods for the determination of selected polychlorinated biphenyl (PCB) congeners, chlorinated pesticides and polybrominated diphenyl ether (PBDE) congeners in human milk and similar matrices.
A unit consists of five bottles of approximately 5 ml fortified human milk.
Zertifizierungen: Traceable to NIST and SRM numbers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge