Potassium standard solution, NIST-3141A
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
NIST-3141A
PROONIST-3141AEA 1410 EUR InStock
PROONIST-3141A
Potassium standard solution, NIST-3141A
Standards Kationischer Standards Kalium-Standards
This reference standard is intended for use as a primary calibration standard for the quantitative determination of potassium.
A unit consists of 50 ml of a single element solution in a high density polyethylene bottle sealed in an aluminised bag.
Zertifizierungen: Traceable to NIST and SRM numbers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge