Sodium carbonate - acidimetric standard
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
NIST-351A
PROONIST-351AEA 0 EUR InStock
PROONIST-351A
Sodium carbonate - acidimetric standard
Standards Kationischer Standards Natrium-Standards
These standards are intended for use as a acidimetric standard.
The SRM consists of highly purified sodium carbonate in a 50 g unit.
Zertifizierungen: Traceable to NIST and SRM.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge