DNPH standards, organotin mixtures, CERTAN®
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SL31005
PROOSL31005EA 607 EUR InStock
PROOSL31005
DNPH standards, organotin mixtures, CERTAN®
Standards Organische Standards
These selected organotin compounds are determined by gas chromatography using DIN 38407-F13.
Zertifizierungen: According to DIN EN ISO 17353.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge