Reference materials, metals in biodiesel Standard, VHG
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VHG-B100M5-5
PROOVHG-B100M5-5EA 242 EUR InStock
PROOVHG-B100M5-5
Reference materials, metals in biodiesel Standard, VHG
Standards Mineralölindustrie-Standards
This solution is intended for use as a calibration or reference standard for the analysis of these analytes in biodiesel or related organic matrices according to ASTM D6751. Sulfur is present in the raw materials and is not certified.
Zertifizierungen: Traceable to NIST, SRM.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge