BOD standard solutions, VHG
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VHG-BOD200A
PROOVHG-BOD200AEA 128 EUR InStock
PROOVHG-BOD200A
BOD standard solutions, VHG
Standards Umweltanalyse-Standards
The standard solution is intended for use as a glucose-glutamic acid standard check solution for biochemical oxygen demand (BOD) in APHA Method 5210 B.
Zertifizierungen: Traceable to NIST.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge