VHG™ Optilene-XF™ XRF Thin Film
Lieferant: LGC Standards PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VHG-FOL06-R3
PROOVHG-FOL06-R3EA 193 EUR InStock
PROOVHG-FOL06-R3
VHG™ Optilene-XF™ XRF Thin Film
Versiegeln Verschlussfolien
Thin films for XRF sample cups.
Highest grade, proprietary film featuring optimal transmission, purity, strength, chemical resistance, and thermal stability. Good for full range of XRF analytes, including light elements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge