Bromate ion chromatography standards, VHG
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VHG-IBRO3-100 VHG-IBRO3-500
PROOVHG-IBRO3-100EA 57 EUR InStock
PROOVHG-IBRO3-100 PROOVHG-IBRO3-500
Bromate ion chromatography standards, VHG
Standards Anionische Standards Bromat-Standards
These standard solutions are intended for use as a calibration or reference standard for ion chromatography (IC) or techniques using other modes of aqueous ion detection.
Zertifizierungen: Traceable to NIST and CRM.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge