SULFUR COMPOUNDS BY SELECTIVE DETECTION FOR ASTM D5623: MULTI-COMPONENT STANDARD CONTAINING 22 SULFUR SPECIES IN BASE FUEL, 2ML AMPO ULE. EACH COMPONENT @ 50G/G. 1 * 2 mL
Lieferant: PROMOCHEM
SULFUR COMPOUNDS BY SELECTIVE DETECTION FOR ASTM D5623: MULTI-COMPONENT STANDARD CONTAINING 22 SULFUR SPECIES IN BASE FUEL, 2ML AMPO ULE. EACH COMPONENT @ 50G/G. 1 * 2 mL
PRODUKT
Erweitert / Kompakt
Beschreibung Katalognummer Verpackungseinheit Menge
SULFUR COMPOUNDS BY SELECTIVE DETECTION FOR ASTM D5623: MULTI-COMPONENT STANDARD CONTAINING 22 SULFUR SPECIES IN BASE FUEL, 2ML AMPO ULE. EACH COMPONENT @ 50µG/G. 1 * 2 mL PROOVHG-MSX22-2 1 * 2 mL
Hersteller-Artikelnummer:: VHG-MSX22-2
Lokale Artikelnummer:: PROOVHG-MSX22-2
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder