Standards for water pollution QC check, total phenolics, VHG
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VHG-QWPPHEN-2
PROOVHG-QWPPHEN-2EA 106 EUR InStock
PROOVHG-QWPPHEN-2
Standards for water pollution QC check, total phenolics, VHG
Standards Umweltanalyse-Standards
This solution is intended for use as a quality control check for methods related to total phenolics.
This standard has been prepared in methanol as a concentrate and must be diluted prior to analysis. This concentrate is not preserved.
Zertifizierungen: Traceable to NIST SRM.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge