Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MADD Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

1150
PRSI1150EA 435 EUR InStock
PRSI1150
Anti-MADD Rabbit Polyclonal Antibody
Antikörper
MADD Antibody: MAP kinase-activating death domain protein (MADD) was initially identified as the type 1 tumor necrosis factor receptor (TNFR1) associated protein though their death domains. Overexpression of MADD activates MAP kinases ERK and JNK and induces the phosphorylation of cytosolic phospholipase A2. MADD shares 98% identity with DENN (for differentially expressed in neoplastic vs. normal cells), which was recently identified as a substrate for c-jun N-terminal kinase 3 (JNK3). MADD has greater than 94% overall identity to a GDP/GTP exchange protein Rab3-GEP. MADD is 87% identical to KIAA0358, a brain protein of unknown function. Identification of MADD as a component of the TNFR1 signaling complex and the similarity between MADD and Rab3-GEP provides a connection between TNFR1 activation and downstream MAP kinase activity through a guanine-nucleotide exchange protein.

Type:
Antigen: MADD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge