Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VPREB1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

25-759
PRSI25-759EA 660 EUR InStock
PRSI25-759
Anti-VPREB1 Rabbit Polyclonal Antibody
Antikörper
VPREB1 belongs to the immunoglobulin superfamily and is expressed selectively at the early stages of B cell development, namely, in proB and early preB cells. This gene encodes the iota polypeptide chain that is associated with the Ig-mu chain to form a molecular complex which is expressed on the surface of pre-B cells. The complex is thought to regulate Ig gene rearrangements in the early steps of B-cell differentiation.CD179a (VpreB) is a 126 aa-long polypeptide with apparent MW of 16-18 kDa. It is expressed selectively at the early stages of B cell development, namely, in proB and early preB cells. CD179a has an Ig V domain-like structure, but lacks the last beta-strand (beta7) of a typical V domain. Instead, it has a carboxyl terminal end that shows no sequence homologies to any other proteins. CD179a associates non-covalently with CD179b (lambda5 or lambda-like) carrying an Ig C domain-like structure to form an Ig light chain-like structure, which is called the surrogate light chain or pseudo light chain. In this complex, the incomplete V domain of CD179a appears to be complemented by the extra beta7 strand of CD179b. On the surface of early preB cells, CD179a/CD179b surrogate light chain is disulfide-linked to membrane-bound Ig mu heavy chain in association with a signal transducer CD79a/CD79b heterodimer to form a B cell receptor-like structure, so-called preB cell receptor (preBCR). Though no CD179a-related human disease or pathology has been reported yet, the deficiency of other components of preB cell receptor such as CD179b, Ig mu heavy chain and CD79a has been shown to result in severe impairment of B cell development and agammaglobulinemia in human. PreBCR transduces signals for: 1) cellular proliferation, differentiation from the proB cell to preB cell stage, 2) allelic exclusion at the Ig heavy chain gene locus, and 3) promotion of Ig light chain gene rearrangements. Thus, preBCR functions as a checkpoint in early B cell development to monitor the production of Ig mu heavy chain through a functional rearrangement of Ig heavy chain gene as well as the potency of Ig mu heavy chain to associate with Ig light chain. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: VPREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge