Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ARL8A Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

26-669
PRSI26-669EA 660 EUR InStock
PRSI26-669
Anti-ARL8A Rabbit Polyclonal Antibody
Antikörper
ARL8A belongs to the small GTPase superfamily, Arf family. ARL8A m,ay play a role in lysosomes motility. Alternatively, may play a role in chromosomes segregation.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ARL8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge