Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SGMS2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

26-739
PRSI26-739EA 660 EUR InStock
PRSI26-739
Anti-SGMS2 Rabbit Polyclonal Antibody
Antikörper
SGMS2 is a bidirectional lipid cholinephosphotransferase capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and vice versa. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. SGMS2 directly and specifically recognizes the choline head group on the substrate. SGMS2 also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. SGMS2 does not function strictly as a SM synthase. SGMS2 is required for cell growth.Sphingomyelin (SM) is a major component of plasma membranes. It is preferentially concentrated in the outer leaflet and has a role in the formation of lipid rafts. SM synthases (EC 2.7.8.27), such as SGMS2, produce SM in the lumen of the Golgi and on the cell surface through the transfer of phosphocholine from phosphatidylcholine onto ceramide, yielding diacylglycerol as a side product (Huitema et al., 2004 [PubMed 14685263]).

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SGMS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge