Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRIP2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

28-067
PRSI28-067EA 660 EUR InStock
PRSI28-067
Anti-CRIP2 Rabbit Polyclonal Antibody
Antikörper
Human ESP1/CRP2 protein has two LIM domains, and each shares 35.1% and 77 or 79% identical residues with human cysteine-rich protein (CRP) and rat CRIP, respectively. Northern blot analysis of ESP1/CRP2 in various human tissues showed distinct tissue distributions compared with CRP and CRIP, suggesting that each might serve related but specific roles in tissue organization or function. Using a panel of human-rodent somatic cell hybrids, the ESP1/CRP2 locus was assigned to chromosome 14. Fluorescence in situ hybridization, using cDNA and a genome DNA fragment of the ESP1/CRP2 as probes, confirms this assignment and relegates regional localization to band 14q32.3.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CRIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge