Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NFIL3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

28-908
PRSI28-908EA 601 EUR InStock
PRSI28-908
Anti-NFIL3 Rabbit Polyclonal Antibody
Antikörper
NFIL3 is a basic leucine zipper (bZIP) transcription factor that represses or activates transcription in non-osteoblastic cells. NFIL3 play a role in attenuation of PTH target gene transcription in osteoblasts. NFIL3 repression domain interacts specifically with the TBP binding repressor protein Dr1.Glucocorticoid-mediated upregulation of the bZIP transcriptional repressor gene, NFIL3, is dependent on [Ca2+]i levels, and correlates with Glucocorticoid-evoked apoptosis of Glucocorticoid-sensitive CEM-C7-14 cells. NFIL3 is involved in a distinct growth factor-regulated signaling pathway that is responsible for the survival of early B-cell progenitors, and whose alteration by E2A-HLF leads to childhood B lineage leukemia.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NFIL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge