Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SAP30BP Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

28-996
PRSI28-996EA 601 EUR InStock
PRSI28-996
Anti-SAP30BP Rabbit Polyclonal Antibody
Antikörper
SAP30BP is a component of a histone deacetylase complex conserved among eukaryotic organisms. This complex is active in deacetylating core histone octamers, but inactive in deacetylating nucleosomal histones due to the inability of the histone binding proteins RbAp46 and RbAp48 to gain access to nucleosomal histones.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SAP30BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge