Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CROT Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

30-463
PRSI30-463EA 601 EUR InStock
PRSI30-463
Anti-CROT Rabbit Polyclonal Antibody
Antikörper
Carnitine octanoyltransferase is a carnitine acyltransferase that catalyzes the reversible transfer of fatty acyl groups between CoA and carnitine. This provides a crucial step in the transport of medium- and long-chain acyl-CoA out of the mammalian peroxisome to the cytosol and mitochondria.Carnitine octanoyltransferase (EC 2.3.1.137) is a carnitine acyltransferase that catalyzes the reversible transfer of fatty acyl groups between CoA and carnitine. This provides a crucial step in the transport of medium- and long-chain acyl-CoA out of the mammalian peroxisome to the cytosol and mitochondria. See also CRAT (MIM 600184). Van der Leij et al. (2000) [PubMed 11001805] reviewed the function, structural features, and phylogenetics of human carnitine acyltransferase genes, including CROT.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CROT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge