Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TIAL1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

31-020
PRSI31-020EA 601 EUR InStock
PRSI31-020
Anti-TIAL1 Rabbit Polyclonal Antibody
Antikörper
The protein encoded by TIAL1 is a member of a family of RNA-binding proteins and possesses nucleolytic activity against cytotoxic lymphocyte target cells. The gene product is a cytotoxic granule-associated protein and has been shown to bind specifically to poly (A) homopolymers and to fragment DNA in permeabilized target cells. It has been suggested that members of this protein family may be involved in the induction of apoptosis. One isoform contains a lysosome-targeting motif in its C-terminal auxiliary domain; however, alternative splicing results in a T-cluster DNA-binding isoform, differing at the C-terminus where a hydrophobic sequence replaces the lysosome-targeting motif.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TIAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge