Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ACP1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-018
PRSI33-018EA 661 EUR InStock
PRSI33-018
Anti-ACP1 Rabbit Polyclonal Antibody
Antikörper
Acid phosphatases are a broad family of enzymes that differ widely in their amino acid length and homology, molecular weight and tissue of origin. They function to remove phosphate groups (or dephosphorylate) from proteins and perform best under acidic conditions. Disease states are sometimes characterised by abnormally high or low levels of Acid Phosphatase, leading to them being used as serological and histological disease markers. Prostatic Acid Phosphatase (PAP) was initially used as a prostate cancer marker but was later replaced with Prostate Serum Antigen (PSA), a more sensitive early stage cancer marker.

  • Western blot: 1 - 3 µg/ml Provided assay concentrations are suggestions only, Acid Phosphatase antibody titration may be required for optimal results.

    1. This antibody is specific to isoforms 2 (or B) and E of Acid Phosphatase.


Type: Primary
Antigen: ACP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge