Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TXNIP Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-035
PRSI33-035EA 661 EUR InStock
PRSI33-035
Anti-TXNIP Rabbit Polyclonal Antibody
Antikörper
TXNIP (Thioredoxin-interacting protein), also called VDUP1 (Vitamin D3 upregulated protein 1) is a member of the Arrestin family of signal transduction proteins. It is a potent tumor suppressor, blocking tumor growth via blocking cell cycle progression, and reduced expression of TXNIP is found in many tumors. One way TXNIP exerts its tumor suppressive function is by suppressing NF-kB activation, possibly via regulation of the NF-kB subunit p65.

Western blot: 1-3 µg/ml Provided assay concentrations are sµggestions only, antibody titration may be required for optimal results.

Type: Primary
Antigen: TXNIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge