Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IPO9 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-040
PRSI33-040EA 661 EUR InStock
PRSI33-040
Anti-IPO9 Rabbit Polyclonal Antibody
Antikörper
Importins transport molecules into the nucleus via nuclear localization sequences binding. There are over 20 human importins identified, divided in the alpha and beta subfamilies. Importin-9 is a beta importin, responsible for the import of the core histones H2A, H2B, H3 and H4 into the nucleus.

Western blot: 0,25-1 µg/ml Provided assay concentrations are sµggestions only, antibody titration may be required for optimal results.

Type: Primary
Antigen: IPO9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge