Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FLT1 Mouse Monoclonal Antibody [clone: FLT1/658]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-133
PRSI33-133EA 773 EUR InStock
PRSI33-133
Anti-FLT1 Mouse Monoclonal Antibody [clone: FLT1/658]
Antikörper
VEGFR1 is high affinity receptors for vascular endothelial growth factors (VEGFs). On the basis of structural similarity to VEGFR1 and 2, it has been speculated that Flt-4 might represent a third receptor for either VEGF or a VEGF-related ligand.

Functional testing: order BSA/azide-free format Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml IF: 1 - 2 µg/ml Variations in protocols, secondaries and substrates may require the antibody to be titered for optimal performance.

Type: Primary
Antigen: FLT1
Clonality: Monoclonal
Clone: FLT1/658
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge