Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ITGAX Mouse Monoclonal Antibody [clone: HC1/1]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-158
PRSI33-158EA 773 EUR InStock
PRSI33-158
Anti-ITGAX Mouse Monoclonal Antibody [clone: HC1/1]
Antikörper
CD11c, also known as Integrin alpha X (complement component 3 receptor 4 subunit) (ITGAX), is a type I transmembrane protein found at high levels on most human dendritic cells, but also on monocytes, macrophages, neutrophils, and some B cells that induces cellular activation and helps trigger neutrophil respiratory burst; expressed in hairy cell leukemias, acute non-lymphocytic leukemias, and some B-cell chronic lymphocytic leukemias.

Flow Cytometry: 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml Functional Activity (Blocking): order BSA/azide free format The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the CD11c antibody to be titered up or down for optimal performance.

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: HC1/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge