Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PAX7 Mouse Monoclonal Antibody [clone: PAX7/497]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-184
PRSI33-184EA 773 EUR InStock
PRSI33-184
Anti-PAX7 Mouse Monoclonal Antibody [clone: PAX7/497]
Antikörper
The PAX gene family of nuclear transcription factors is comprised of nine members that function during embryogenesis to regulate the temporal and position-dependent differentiation of cells. In addition, the family is involved in a variety of signal transduction pathways in the adult organism. Mutations in the PAX family of proteins have been linked to disease and cancer in humans. PAX7 is a protein specifically expressed in cultured satellite cell- derived myoblasts. In situ hybridization reveals that PAX7 is also expressed in satellite cells residing in adult muscle. A chromosomal aberration in the gene encoding PAX7 causes rhabdomyosarcoma 2 (RMS2) (also called alveolar rhabdomyosarcoma).

Flow Cytometry: 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml WB: 0,5 - 1 µg/ml The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the antibody to be titered up or down for optimal performance.

Type: Primary
Antigen: PAX7
Clonality: Monoclonal
Clone: PAX7/497
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse, Rat Chicken, Zebrafish

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge