Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CK4, CK5, 6, 8, 10, 13, and 18 Mouse Monoclonal Antibody [clone: C11]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-241
PRSI33-241EA 773 EUR InStock
PRSI33-241
Anti-CK4, CK5, 6, 8, 10, 13, and 18 Mouse Monoclonal Antibody [clone: C11]
Antikörper
This antibody recognises multi cytokeratins: 4, 5, 6, 8, 10, 13, and 18. It is a broad-spectrum cytokeratin antibody which has been reported to be used to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of cytokeratin markers in cancer research and tumor diagnosis.

Flow Cytometry: use the Multi Cytokeratin mAb at 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml WB: 0,5 - 1 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Multi Cytokeratin antibody to be titered up or down for optimal performance.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: Multi Cytokeratin 4/5/6/8/10/13/18 Antibody [C11]
Clonality: Monoclonal
Clone: C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Rat, Mouse, Guinea pig, Frog, Goat, Marmoset, Pig

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge