Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD14 Mouse Monoclonal Antibody [clone: LPSR/654]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-303
PRSI33-303EA 0 EUR InStock
PRSI33-303
Anti-CD14 Mouse Monoclonal Antibody [clone: LPSR/654]
Antikörper
Recognises a protein of 55 kDa, identified as Cluster of Differentiation 14 (also known lipopolysaccharide receptor or LPSR). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. It is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.

Flow Cytometry: 0,5-1 µg/million cells Immunofluorescence: 0,5 - 1 µg/ml Variations in protocols, secondaries and substrates may require the CD14 antibody to be titered up or down for optimal performance.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: LPSR/654
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge