Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CNN1 Mouse Monoclonal Antibody [clone: CNN1/832]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-336
PRSI33-336EA 773 EUR InStock
PRSI33-336
Anti-CNN1 Mouse Monoclonal Antibody [clone: CNN1/832]
Antikörper
Multiple isoelectric variants of calponin have been identified, however only two molecular weight isoforms exist; a 34 kDa form and a 29 kDa form. Expression of the 29 kDa form, I-calponin, is primarily restricted to muscle of the urogenital tract, whereas the higher molecular weight variant has been demonstrated in vascular and visceral smooth muscle. Calponin is a calmodulin, F-actin and tropomyosin binding protein, which is thought to be involved in the regulation of smooth muscle contraction. Calponin expression is restricted to smooth muscle cells and has been shown to be a marker of the differentiated (contractile) phenotype of developing smooth muscle.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the anti-Calponin antibody should be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Tris with 1mM EDTA, pH 9.0, for 10 - 20 min followed by cooling at RT for 20 min
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: CNN1
Clonality: Monoclonal
Clone: CNN1/832
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge