Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IFNA2 Mouse Monoclonal Antibody [clone: N39]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-405
PRSI33-405EA 773 EUR InStock
PRSI33-405
Anti-IFNA2 Mouse Monoclonal Antibody [clone: N39]
Antikörper
Recognises a protein of 16-27 kDa, identified as human interferon-II) (IFN-(II). Its epitope maps between aa112-148 of IFN-II) (total aa172). This mAb is specific for IFN-(II) and does not cross-react with IFN-(I). The site recognized by this mAb is called "site I" and is responsible for the antiviral and anti-proliferative activities of IFN-(II). Epitopes of N27 and N39 mAbs are different and represent a good combination of antibodies to set up an ELISA assay for the quantitation of IFN-(II) after viral infections. The IFN- family consists of 24 or more genes or pseudo-genes. IFN-II) is one of the two distinct families (I and II) of human IFN-. The -interferon are mainly produced by lymphocytes, monocytes, macrophages, and cell lines such as Namalwa and KG1 following induction by viruses, nucleic acids, and glucocorticoid hormones. They are involved in virus resistance on target cells, inhibition of cell proliferation, induction of cytokines and regulation of expression of MHC class I antigens.

ELISA: order BSA/azide-free format for coating Optimal dilution of the Interferon alpha 2 antibody should be determined by the researcher.

Type: Primary
Antigen: IFNA2
Clonality: Monoclonal
Clone: N39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG, kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge