Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FITC Mouse Monoclonal Antibody [clone: SPM395]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-709
PRSI33-709EA 773 EUR InStock
PRSI33-709
Anti-FITC Mouse Monoclonal Antibody [clone: SPM395]
Antikörper
This mAb recognises both the free and protein-conjugated (either soluble or cell bound) form of fluorescein or carboxy-fluorescein. Sensitivity of the FITC-anti-FITC system is reportedly similar to that of the biotin-avidin system. In biotin or enzyme-labeled form, this mAb is also suitable for the conversion of fluorescein-labeled detection into immuno-enzymatic assays.

Confocal Microscopy: suitable Flow Cytometry: 0,5 - 1 µg/10^6 cells in 0,1 ml IF: 0,5 - 1 µg/ml WB: 0,5 - 1 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT Optimal dilution of the FITC antibody should be determined by the researcher.

Type: Primary
Antigen: FITC Antibody [SPM395]
Clonality: Monoclonal
Clone: SPM395
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge