Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Mitochondrial antigen Mouse Monoclonal Antibody [clone: MTL56]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-878
PRSI33-878EA 773 EUR InStock
PRSI33-878
Anti-Mitochondrial antigen Mouse Monoclonal Antibody [clone: MTL56]
Antikörper
The mitochondrial marker antibody is specific for an antigen associated with the mitochondria in human cells. It can be used to stain mitochondria in cell or tissue preparations and as a mitochondrial marker in subcellular fractions.

IHC (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) IF: 0,5 - 1 µg/ml Western blot: 1 - 2 µg/ml Titering of the Mitochondrial antibody may be required for optimal performance.1. FFPE staining is enhanced by boiling tissue sections in 10 mM citrate buffer, pH 6, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: Mitochondrial Antibody [MTL56]
Clonality: Monoclonal
Clone: MTL56
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge