Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-WT1 Mouse Monoclonal Antibody [clone: WLMT1-1]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

33-996
PRSI33-996EA 773 EUR InStock
PRSI33-996
Anti-WT1 Mouse Monoclonal Antibody [clone: WLMT1-1]
Antikörper
Wilms Tumor is a transcription factor that plays an important role in cellular development and cell survival. Regulates the expression of numerous target genes, including EPO. Plays an essential role for development of the urogenital system. recognises and binds to the DNA sequence 5'-CGCCCCCGC-3'. It has a tumor suppressor as well as an oncogenic role in tumor formation. Function may be isoform-specific: isoforms lacking the KTS motif may act as transcription factors. Isoforms containing the KTS motif may bind mRNA and play a role in mRNA metabolism or splicing. Isoform 1 has lower affinity for DNA, and can bind RNA. [UniProt].

Immunofluorescence: 1 - 2 µg/ml Immunohistochemistry (FFPE): 1 - 2 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Titering of the WT1 antibody may be required for optimal performance.

1. FFPE testing requires sections to be boiled in pH6 10 mM citrate buffer for 10 - 20 minutes, followed by cooling at RT for 20 minutes, prior to staining.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: WT1
Clonality: Monoclonal
Clone: WLMT1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge