Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD1A Rabbit Monoclonal Antibody [clone: C1A/1506R]

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

34-051
PRSI34-051EA 773 EUR InStock
PRSI34-051
Anti-CD1A Rabbit Monoclonal Antibody [clone: C1A/1506R]
Antikörper
CD1a is a non-polymorphic MHC Class 1 related cell surface glycoprotein expressed in association with Beta-2 microglobulin.

Flow Cytometry: 0,5-1 µg/million cells IF: 1 - 2 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT Prediluted IHC only format: incubate for 30 min at RT (1) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the recombinant CD1a antibody to be titered up or down for optimal performance.

1. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: C1A/1506R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG, kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge