Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Canine Heartworm Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

35-127
PRSI35-127EA 549 EUR InStock
PRSI35-127
Anti-Canine Heartworm Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Purified IgG fraction of antiserum covalently coupled with highly purified preparation of horseradish peroxidase (RZ>3). Care is taken to insure adequate conjugation while preserving the maximum enzyme activity. Free enzyme is absent.

Canine Heartworm antibody is in a phosphate saline buffer with BSA (10mg/ml) and preserved with 0.002% thimerosal.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Canine Heartworm
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Goat
Isotype:
Reactivity: Parasite

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge