Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PTEN Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

3517
PRSI3517EA 435 EUR InStock
PRSI3517
Anti-PTEN Rabbit Polyclonal Antibody
Antikörper
PTEN Antibody: PTEN (phosphatase and tensin homologue deleted on chromosome ten) is a dual-specificity phosphatase (with both protein and lipid phosphatase activity) first identified as a tumor suppressor gene. PTEN indirectly activates the AKT/PI3K pathway, an important signaling pathway for cell growth and proliferation by keeping levels of the second messenger PIP3 low, thereby preventing phosphoinositide-dependent kinase-1 (PDK-1) from phosphorylating and activating AKT. Expression of wild-type PTEN causes growth arrest in many cancer cell lines, but expression of a PTEN protein containing a mutation that blocks its lipid phosphatase activity failed to suppress growth of glioma cell lines suggesting that the tumor suppressive effect of PTEN is mediated solely by its lipid phosphatase activity. Other activities include the inhibition of insulin stimulated MAPK activation by blocking the insulin-receptor substrate (IRS)-1 phosphorylation and assembly of the IRS-1/Grb2/Sos complex.

Type:
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge