Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FEZ1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

4265
PRSI4265EA 435 EUR InStock
PRSI4265
Anti-FEZ1 Rabbit Polyclonal Antibody
Antikörper
FEZ1 Antibody: Similar to its C. elegans homolog UNC-76, mammalian FEZ1, an abundant membrane protein and protein kinase C (PKC)-Z substrate, is involved in axonal guidance. Following phosphorylation by PKC-Z, FEZ1 translocates from the plasma membrane to the cytoplasm. FEZ1 is then able to interact with a number of proteins involved in axonal outgrowth and cellular transport such as DISC1, a candidate gene for schizophrenia, as well as kinesin-1, a microtubule-based motor protein. FEZ1 also interacts with proteins involved in transcriptional regulation and chromatin remodeling, suggesting that FEZ1 may have nuclear regulatory functions in addition to those for neuronal development. At least two distinct isoforms of FEZ1 are known to exist.

Type:
Antigen: FEZ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge