Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CHRNB1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

46-824
PRSI46-824EA 620 EUR InStock
PRSI46-824
Anti-CHRNB1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 55 kDa band observed in human brain (cerebellum, hippocampus, frontal cortex) lysates (calculated MW of 56.7 kDa according to NP_000738.2). Recommended concentration: 1-3 μg

CHRNB1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CHRNB1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHRNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge