Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RB1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

47-180
PRSI47-180EA 620 EUR InStock
PRSI47-180
Anti-RB1 Rabbit Polyclonal Antibody
Antikörper
Epithelium-specific gene regulation plays acritical role during embryogenesis, and the epithelial cell lineage is the first differentiated cell type to appear afterfertilization. Until recently, very few epithelial cell-restricted transcription factors had been described. Members of the ets transcription factor/oncogene family havebeen implicated in human carcinogenesis following the discovery of several distinctchromosomal translocations involving members of the ets family in different types of cancer. ets factors play key roles intranscriptional control of stringently regulated genes, such as genes involved intissue development, differentiation,angiogenesis, cell cycle control and cellproliferation. ESE-1 is the first member of the ets family that was found to be exclusively expressed and inducible inepithelial cells.

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge