Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ACVR1B Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

4793
PRSI4793EA 435 EUR InStock
PRSI4793
Anti-ACVR1B Rabbit Polyclonal Antibody
Antikörper
ACVR1B Antibody: Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I and two type II receptors. ACVR1B, also known as activin receptor-like kinase 4 (ALK4), is a type I receptor for activin and plays major roles in cell differentiation, growth arrest and apoptosis. Like another type I activin receptor ACVR1C, ACVR1B can mediate signaling by ligands such as Nodal, Xnr1, GDF-1/3, activin B and activin AB. In Xenopus embryos, expression of a dominant-negative form of ACVR1B blocked all mesoderm-inducing ligands, while expression of a dominant negative ACVR1C only blocked Nodal and Xnr1 signaling, suggesting that the ACVR1B and ACVR1C possess distinct functions.

Type:
Antigen: ACVR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge