Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ABCB1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

48-673
PRSI48-673EA 815 EUR InStock
PRSI48-673
Anti-ABCB1 Rabbit Polyclonal Antibody
Antikörper
ATP-Binding Cassette Sub-Family B Member 1 (ABCB1, also named P-glycoprotein) is a plasma membrane-associated multidrug transporter that utilizes the energy of ATP hydrolysis to pump toxic xenobiotics out of cells. Unique features of ABCB1 are its very broad substrate specificity and its basal ATPase activity in the absence of transport substrates. Human ABCB1 plays an important role in absorption, distribution, metabolism, excretion and toxicity of pharmacologically relevant drugs. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anti-cancer drugs. This protein also functions as a transporter across the blood-brain barrier.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ABCB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Monkey, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge