Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TGFB1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

49-553
PRSI49-553EA 815 EUR InStock
PRSI49-553
Anti-TGFB1 Rabbit Polyclonal Antibody
Antikörper
TGF-beta-1 is a multifunctional cytokine that belongs to a superfamily of structurally related regulatory proteins, which includes three mammalian TGF-beta isoforms (TGF-beta-1, -beta-2, and -beta-3), activin/inhibins and bone morphogenetic proteins. The most abundant isoform, TGF-beta-1, is a 25 kDa homodimer composed of two 12.5 kDa subunits joined by disulfide bonds. TGF-beta-1 is a highly conserved molecule - the amino acid sequence between human and mouse differ only by one residue. Although originally defined by its ability to cause anchorage independent cell growth and changes in cell morphology of rat fibroblasts, subsequent research has revealed that TGF-beta is actually a major growth inhibitor for most cell types. It is produced by a wide variety of cell and tissue types during all stages of cell differentiation. TGF-beta-1 sources include platelets, bone and soft tissues such as placenta and kidneys.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TGFB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge