Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MIPU1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

5219
PRSI5219EA 435 EUR InStock
PRSI5219
Anti-MIPU1 Rabbit Polyclonal Antibody
Antikörper
MIPU1 Antibody: Mipu1, also known as zinc finger protein 667 or ZNF667, encodes a nuclear-localized protein containing 14 carboxy-terminal zinc finger motifs and an amino-terminal KRAB domain. This protein is highly expressed in heart and brain and is upregulated in rat heart after a transient ischemia-reperfusion procedure. Overexpression experiments suggest that Mipu1 suppresses the transcriptional activities of AP-1 and SRE in the MAPK signaling pathway and thus may play a role in the pathogenesis of cardiac and vascular disease. At least four isoforms of MIPU1 are known to exist.

Type:
Antigen: MIPU1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge